Learn More
MSC Ringers Gloves R169 Impact Gloves, Synthetic Leather, HPPE, 3X-Large, 5/Pk.
Catalog No. 5000687605
Encompass
These medium-duty impact gloves utilize F3 Technology™ and a thermoplastic rubber (TPR) design to protect the knuckles, thumbs, and fingers. Constructed with a durable HPPE liner and a premium synthetic leather palm, these gloves provide robust cut and puncture resistance for demanding industrial applications.
  • Provides ANSI Cut Level A5 and ANSI Puncture Level 3 protection
  • Features impact protection compliant with ANSI/ISEA 138-2019 Level 2
  • Constructed with a premium synthetic leather palm for enhanced durability
  • Incorporates a hook and loop rubber pull tab for a secure, adaptable fit
  • Designed with a seamless index finger to minimize wear in high-stress areas
  • Suitable for manufacturing, mining, oil and gas, and heavy machinery operations
  • Machine washable at 30 °C (86 °F)
Catalog No. 50-006-87605 Supplier MSC Supplier No. 58717372
These medium-duty impact gloves utilize F3 Technology™ and a thermoplastic rubber (TPR) design to protect the knuckles, thumbs, and fingers. Constructed with a durable HPPE liner and a premium synthetic leather palm, these gloves provide robust cut and puncture resistance for demanding industrial applications.
  • Provides ANSI Cut Level A5 and ANSI Puncture Level 3 protection
  • Features impact protection compliant with ANSI/ISEA 138-2019 Level 2
  • Constructed with a premium synthetic leather palm for enhanced durability
  • Incorporates a hook and loop rubber pull tab for a secure, adaptable fit
  • Designed with a seamless index finger to minimize wear in high-stress areas
  • Suitable for manufacturing, mining, oil and gas, and heavy machinery operations
  • Machine washable at 30 °C (86 °F)