Learn More
New England Biolabs, Inc. IMPACT™ KIT – 1 set
SDP
Catalog No. 50994917
Encompass
The IMPACT (Intein Mediated Purification with an Affinity Chitin-binding Tag) system is a novel protein purification system which utilizes the inducible self-cleavage activity of protein splicing elements, termed inteins, to separate the target protein from the affinity tag
Catalog No. 50-994-917 Supplier New England Biolabs, Inc. Supplier No. E6901S
The IMPACT (Intein Mediated Purification with an Affinity Chitin-binding Tag) system is a novel protein purification system which utilizes the inducible self-cleavage activity of protein splicing elements, termed inteins, to separate the target protein from the affinity tag