Learn More
Metrohm USA IMPACT PRINTER DP40-S4N
SDP
Supplier:  Metrohm USA 21400200
Encompass
  • Impact printer with reliable data output
  • Normal paper type for immediate hardcopy documentation
  • Supports 58 mm paper width
  • Provides 40 columns of text
  • Includes a serial RS 232C (DB9) interface for easy integration
  • Operates on a universal power supply supporting 100-240 V
  • Compatible with 50/60 Hz frequency
Catalog No. NC1070057