Learn More
Enterprise Technology Solutions Impact Products Cleaning Cloths, Microfiber, 16"x16", 12/BG, Yellow
SDP
Catalog No. 5000831614
Encompass_Preferred
Premium-weight microfiber cloths are a great choice for dusting and cleaning. Split fibers create a positive charge that grasps dust and dirt without damaging surfaces. Microfiber is safe for dry dusting hardwoods and for dusting and cleaning without chemicals or oils. Microfiber dusters have the same great qualities as cloths but can be used in harder-to-reach areas like ceiling fans, shelves and irregular surfaces. Microfiber cloth is perfect for washroom cleaning and general-purpose cleaning. It can be extensively laundered.
Catalog No. 50-008-31614 Supplier Enterprise Technology Solutions Supplier No. IMPLFK700
Premium-weight microfiber cloths are a great choice for dusting and cleaning. Split fibers create a positive charge that grasps dust and dirt without damaging surfaces. Microfiber is safe for dry dusting hardwoods and for dusting and cleaning without chemicals or oils. Microfiber dusters have the same great qualities as cloths but can be used in harder-to-reach areas like ceiling fans, shelves and irregular surfaces. Microfiber cloth is perfect for washroom cleaning and general-purpose cleaning. It can be extensively laundered.