Learn More
Enterprise Technology Solutions Impact Products Dispensing Pump,38/400mm Thread,11-1/2" Tube,White
SDP
Catalog No. 5000810881
Encompass_Preferred
Use this 4 cc Dispensing Pump for easier application of hand sanitizers, hand soaps, lotions and more out of larger containers (not included). Its 38/400mm thread finish is compatible with most 1 gallon and 1/2 gallon jugs. The 4 cc output is ideal for use with liquid and gel hand sanitizers. The pump tube is 11-1/2" long.
Catalog No. 50-008-10881 Supplier Enterprise Technology Solutions Supplier No. IMP940
Use this 4 cc Dispensing Pump for easier application of hand sanitizers, hand soaps, lotions and more out of larger containers (not included). Its 38/400mm thread finish is compatible with most 1 gallon and 1/2 gallon jugs. The 4 cc output is ideal for use with liquid and gel hand sanitizers. The pump tube is 11-1/2" long.