Learn More
Enterprise Technology Solutions Impact Products Fill Container, 5 Gal, 9-1/2"Wx16"Dx9-1/2"H, Translucent
SDP
Catalog No. 5000831688
Encompass_Preferred
E-Z Fill Container offers a user-friendly solution for easy filling and pouring. Use the 63mm angled front opening on this heavy-duty container to dispense ready-to-use chemical concentrates. 38mm top vent opening accepts standard 3/4" tubing. Container includes a 7577 faucet with 38mm cap no-leak design. Spout fits into 28mm bottle neck so no spilling. Measuring is easy with 1/2-gallon embossed graduations on top and front. U.S. patented container is FDA-compliant.
Catalog No. 50-008-31688 Supplier Enterprise Technology Solutions Supplier No. IMP7576
E-Z Fill Container offers a user-friendly solution for easy filling and pouring. Use the 63mm angled front opening on this heavy-duty container to dispense ready-to-use chemical concentrates. 38mm top vent opening accepts standard 3/4" tubing. Container includes a 7577 faucet with 38mm cap no-leak design. Spout fits into 28mm bottle neck so no spilling. Measuring is easy with 1/2-gallon embossed graduations on top and front. U.S. patented container is FDA-compliant.