Learn More
Enterprise Technology Solutions Impact Products Mophead, Wet, Microfiber, Flat, 5"Wx18"L, 12/BG, Blue
SDP
Catalog No. 5000831674
Encompass_Preferred
Microfiber wet mophead features polypropylene scrubbing strips that sit vertically for excellent, deep grout-line cleaning. An additional layer of microfiber enhances its fluid retention vs. a layer of foam, making it more bacteria-resistant. Durable design is washable to maintain cleanliness. Mophead is designed for Impact hook-and-loop-style microfiber mop frames (sold separately).
Catalog No. 50-008-31674 Supplier Enterprise Technology Solutions Supplier No. IMPLWBS18
Microfiber wet mophead features polypropylene scrubbing strips that sit vertically for excellent, deep grout-line cleaning. An additional layer of microfiber enhances its fluid retention vs. a layer of foam, making it more bacteria-resistant. Durable design is washable to maintain cleanliness. Mophead is designed for Impact hook-and-loop-style microfiber mop frames (sold separately).