Learn More
Enterprise Technology Solutions Impact Products Plastic Bottles w/Graduation, 32oz., Frosted
SDP
Catalog No. 5000831753
Encompass_Preferred
Plastic bottles offer embossed graduations for easy measuring. Each bottle is made of polyethylene and is FDA-compliant. The 28mm/400 neck finish fits a variety of popular trigger sprayers. Bottle also includes dilution ratios marked on it.
Catalog No. 50-008-31753 Supplier Enterprise Technology Solutions Supplier No. IMP5032WG
Plastic bottles offer embossed graduations for easy measuring. Each bottle is made of polyethylene and is FDA-compliant. The 28mm/400 neck finish fits a variety of popular trigger sprayers. Bottle also includes dilution ratios marked on it.