Learn More
Enterprise Technology Solutions Impact Products Sharps Container, 2Gal Capacity, Red
SDP
Supplier:  Enterprise Technology Solutions IMP7352
Encompass_Preferred
Sharps Container with 2 gallon capacity meets or exceeds OSHA, NIOSH, CDC, and JCAHO Standards. Puncture-resistant design exceeds current ASTM standards for wall thickness. Latex-free and heavy-metal free for added safety, this container is recommended for use in patients' rooms, physicians' offices, clinics and other high-traffic areas. Lid design limits access to container contents with a lock for disposal. Built-in needle key allows safe removal of needles when necessary. Horizontal wide drop allows maximum capacity of sharps. It can accommodate syringes up to 60cc. Translucent container and lid help identify fill level. Container is nestable to save money in shipping and storage. Built-in handles allow easy transport.
Catalog No. 50-008-31091