Learn More
Enterprise Technology Solutions Impact Products Siphon Drum Pump, Polyethylene, 3"W x 45"L, Red/White
SDP
Catalog No. 5000811297
Encompass_Preferred
Siphon drum pump lets you efficiently transfer liquids where needed. Polyethylene construction is durable for long-lasting use. Advanced design siphons 3.6 gallons per minute. Drum pump fits 15, 30 and 55-gallon drums with a 2" National Pipe Thread.
Catalog No. 50-008-11297 Supplier Enterprise Technology Solutions Supplier No. IMP2300
Siphon drum pump lets you efficiently transfer liquids where needed. Polyethylene construction is durable for long-lasting use. Advanced design siphons 3.6 gallons per minute. Drum pump fits 15, 30 and 55-gallon drums with a 2" National Pipe Thread.