Learn More
Enterprise Technology Solutions Impact Products Sprayer, Pump Up, 2 Liter Tank, 5-3/4"Wx5-1/4"Lx12-1/5"H, BK
SDP
Catalog No. 5000831616
Encompass_Preferred
Pump-up sprayer with a 2-liter tank capacity features an ergonomic thumb-lock that makes it easier to spray for fatigue-free spraying. Included foam tip provides excellent foam generation for multipurpose performance. 70 PSI output at full compression provides plenty of power to help you spray hard-to-reach locations. High-density polypropylene prevents bending or crushing for lasting use in rough environments.
Catalog No. 50-008-31616 Supplier Enterprise Technology Solutions Supplier No. IMP6500
Pump-up sprayer with a 2-liter tank capacity features an ergonomic thumb-lock that makes it easier to spray for fatigue-free spraying. Included foam tip provides excellent foam generation for multipurpose performance. 70 PSI output at full compression provides plenty of power to help you spray hard-to-reach locations. High-density polypropylene prevents bending or crushing for lasting use in rough environments.