Learn More
Grainger Impact Wrench 258 ft.-lb. Max Torque
Catalog No. 502749191
Encompass_Preferred
The Grainger 14F191 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with 3/8-inch drive ratchets, handles, extensions, etc. It measures 1-1/2 inches long and weighs 0.8 ounces. The adapter has a torque rating of 250 inch-pounds.
Catalog No. 50-274-9191 Supplier Grainger Supplier No. 14F191
The Grainger 14F191 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with 3/8-inch drive ratchets, handles, extensions, etc. It measures 1-1/2 inches long and weighs 0.8 ounces. The adapter has a torque rating of 250 inch-pounds.