Learn More
Grainger Proto Impact Universal Joint, 1 in. Drive, Black Oxide, 1 ea.
Catalog No. 502792572
Encompass_Preferred
This 1 in. drive impact universal joint is designed for use with powered drive tools to install and remove fasteners in applications where obstructions prevent a straight-line approach. Constructed from durable alloy steel with a black oxide finish, it is engineered to withstand the high torque and forces associated with repetitive impact tool usage.
  • Features a ball design that allows for a 30° working angle and smooth 360° rotation.
  • Includes two through holes and a groove to accept a retaining ring or pin and O-ring to secure the socket to the drive tool anvil.
  • Designed with tight tolerances for a snug fit on fasteners.
  • Meets ASME B107.110-2012 standards.
  • Measures 4-19/32 in. overall length.
Catalog No. 50-279-2572 Supplier Grainger Supplier No. 1ACR9
This 1 in. drive impact universal joint is designed for use with powered drive tools to install and remove fasteners in applications where obstructions prevent a straight-line approach. Constructed from durable alloy steel with a black oxide finish, it is engineered to withstand the high torque and forces associated with repetitive impact tool usage.
  • Features a ball design that allows for a 30° working angle and smooth 360° rotation.
  • Includes two through holes and a groove to accept a retaining ring or pin and O-ring to secure the socket to the drive tool anvil.
  • Designed with tight tolerances for a snug fit on fasteners.
  • Meets ASME B107.110-2012 standards.
  • Measures 4-19/32 in. overall length.