Learn More
Medchemexpress LLC Impdh-in-3 | 401822-29-5 | 90.0% | 10 MG
SDP
Catalog No. 5000342886
Encompass_Preferred
IMPDH-IN-3 (Compound C67) is an inhibitor of *C. parvum* IMPDH (CpIMPDH) with an IC50 of 35 nM. It can be used in the research of anti-Cryptosporidium drugs.
  • Inhibits C. parvum IMPDH
  • Has an IC50 of 35 nM
  • Useful in the research of anti-Cryptosporidium drugs
Catalog No. 50-003-42886 Supplier Medchemexpress LLC Supplier No. HY12821410MG
IMPDH-IN-3 (Compound C67) is an inhibitor of *C. parvum* IMPDH (CpIMPDH) with an IC50 of 35 nM. It can be used in the research of anti-Cryptosporidium drugs.
  • Inhibits C. parvum IMPDH
  • Has an IC50 of 35 nM
  • Useful in the research of anti-Cryptosporidium drugs