Learn More
Aviva Systems Biology Corp IMPDH2 Antibody - C-terminal region (ARP54366_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54366_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IMPDH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human IMPDH2. Alternate names: IMPD2, IMPDH-IIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61187