Learn More
Cole-Parmer Lightnin 808955 High-Efficiency Axial Flow Impeller, 5/16" Bore; 2.5" Dia
Catalog No. NC9507747 Shop All Cole-Parmer Products
Encompass_Preferred
High-efficiency axial flow impellers have laser designed paddles to develop 50% more mixing action for high-flow, low-shear mixing.
This item is not returnable. View return policy
Catalog No. NC9507747 Supplier Cole-Parmer Supplier No. 0456025
High-efficiency axial flow impellers have laser designed paddles to develop 50% more mixing action for high-flow, low-shear mixing.