Learn More
Bellco Glass, Inc. Impeller Assembly ,15-36L
SDP
Catalog No. 501945153
Encompass_Preferred
Glass shaft impeller assembly for 1967-style hanging bar spinner flask. Consist of ptfe magnet holder and ptfe coated magnet. MADE IN THE USA. Glass shaft ptfe magnet Size: 15-36l CONSIST OF: PTFE MAGNET HOLDER and PTFE COATED MAGNET
Catalog No. 50-194-5153 Supplier Bellco Glass, Inc. Supplier No. 196931500
Glass shaft impeller assembly for 1967-style hanging bar spinner flask. Consist of ptfe magnet holder and ptfe coated magnet. MADE IN THE USA. Glass shaft ptfe magnet Size: 15-36l CONSIST OF: PTFE MAGNET HOLDER and PTFE COATED MAGNET