Learn More
Bellco Glass, Inc. Impeller Assembly ,1-3L
SDP
Catalog No. 501945152
Encompass_Preferred
Glass shaft impeller assembly for 1967-style hanging bar spinner flask. Consist of ptfe magnet holder and ptfe coated magnet. MADE IN THE USA. Glass shaft ptfe magnet Size: 1-3l CONSIST OF: PTFE MAGNET HOLDER and PTFE COATED MAGNET
Catalog No. 50-194-5152 Supplier Bellco Glass, Inc. Supplier No. 196931000
Glass shaft impeller assembly for 1967-style hanging bar spinner flask. Consist of ptfe magnet holder and ptfe coated magnet. MADE IN THE USA. Glass shaft ptfe magnet Size: 1-3l CONSIST OF: PTFE MAGNET HOLDER and PTFE COATED MAGNET