Learn More
Bellco Glass, Inc. Impeller Assembly , 25mL
SDP
Catalog No. 501945148
Encompass_Preferred
Glass shaft impeller assembly for 1967-style hanging bar spinner flask. Consist of ptfe magnet holder and ptfe coated magnet. MADE IN THE USA. Glass shaft ptfe magnet Size: 25ml
Catalog No. 50-194-5148 Supplier Bellco Glass, Inc. Supplier No. 196730025
Glass shaft impeller assembly for 1967-style hanging bar spinner flask. Consist of ptfe magnet holder and ptfe coated magnet. MADE IN THE USA. Glass shaft ptfe magnet Size: 25ml