Learn More
Chemglass Life Sciences PTFE Impeller Assembly, 241mm OAW x 442mm OAH, for 36L VSA
SDP
Catalog No. 50143202
Encompass
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.
Catalog No. 50-143-202 Supplier Chemglass Life Sciences Supplier No. CLS144030
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.