Learn More
Chemglass Life Sciences PTFE Impeller Assembly, 108mm OAW x 222mm OAH, for 3L Spinner
SDP
Catalog No. 50143165
Encompass
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.
Catalog No. 50-143-165 Supplier Chemglass Life Sciences Supplier No. CLS144026
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.