Learn More
Chemglass Life Sciences PTFE Impeller Assembly, 171mm OAW x 355mm OAH, for 6L Spinner
SDP
Catalog No. 50143222
Encompass
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.
Catalog No. 50-143-222 Supplier Chemglass Life Sciences Supplier No. CLS144027
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.