Learn More
Chemglass Life Sciences SST Pitch Blade Impeller, Low Profile 6L VSA, 6.5" W x 2.5" H Blade
SDP
Catalog No. 50143707
Encompass
Our SS pitch blade folding impeller assemblies are designed to provide optimum agitation forVertical Side Arm (VSA) vessels.These impellers will improve uplift mixing and increase mass transfer rates while minimizing shear to cultures.They can be used withcap adapter and Dura Drive Overhead Motor.The impellers are made from 316 SS and supplied with a dust shield as well as a locking ring to secure the paddle in a horizontal position once inserted into the flask.
Catalog No. 50-143-707 Supplier Chemglass Life Sciences Supplier No. CLS1440P53
Our SS pitch blade folding impeller assemblies are designed to provide optimum agitation forVertical Side Arm (VSA) vessels.These impellers will improve uplift mixing and increase mass transfer rates while minimizing shear to cultures.They can be used withcap adapter and Dura Drive Overhead Motor.The impellers are made from 316 SS and supplied with a dust shield as well as a locking ring to secure the paddle in a horizontal position once inserted into the flask.