Learn More
Chemglass Life Sciences SST Pitch Blade Impeller, 15L VSA, 6.5" W x 2.5" H Blade
SDP
Catalog No. 50143795
Encompass
Our SS pitch blade folding impeller assemblies are designed to provide optimum agitation forVertical Side Arm (VSA) vessels.These impellers will improve uplift mixing and increase mass transfer rates while minimizing shear to cultures.They can be used withcap adapter and Dura Drive Overhead Motor.The impellers are made from 316 SS and supplied with a dust shield as well as a locking ring to secure the paddle in a horizontal position once inserted into the flask.
Catalog No. 50-143-795 Supplier Chemglass Life Sciences Supplier No. CLS1440P49
Our SS pitch blade folding impeller assemblies are designed to provide optimum agitation forVertical Side Arm (VSA) vessels.These impellers will improve uplift mixing and increase mass transfer rates while minimizing shear to cultures.They can be used withcap adapter and Dura Drive Overhead Motor.The impellers are made from 316 SS and supplied with a dust shield as well as a locking ring to secure the paddle in a horizontal position once inserted into the flask.