Learn More
Chemglass Life Sciences 1L Internal Large Impeller Spinner Flask, Bioprocess, Complete
SDP
Catalog No. 501311273
Encompass_Preferred
MicroCarrier spinner flasks with larger paddle design promotes increased optimal growth of suspension and microcarrier cultures.Indented bottom (100mL thru 1000mL) promotes mixing action under the impeller paddle.The 3L version does not have a dimple.The complete system includes a flask with two angled sidearms and a PTFE paddle assembly that attaches underneath the cap decreasing the risk of contamination.
Catalog No. 50-131-1273 Supplier Chemglass Life Sciences Supplier No. CLS14551L
MicroCarrier spinner flasks with larger paddle design promotes increased optimal growth of suspension and microcarrier cultures.Indented bottom (100mL thru 1000mL) promotes mixing action under the impeller paddle.The 3L version does not have a dimple.The complete system includes a flask with two angled sidearms and a PTFE paddle assembly that attaches underneath the cap decreasing the risk of contamination.