Learn More
AdipoGen Imperatorin (25 mg)
SDP
Supplier:  AdipoGen AGCN20084M025
Encompass_Preferred
Chemical. CAS: 482-44-0. Formula: C16H14O4. MW: 270.3. Isolated from Opopanax chironium L. Anticancer compound. Apoptosis inducer. Induces DNA fragmentation at the G1/S phase of the cell cycle. HIV-1 replication inhibitor. Anti-inflammatory. Vasodilatation inducer. Increases BMP-2 expression (mRNA) and enhance bone formation. Modulator of SMAD1/5/8, p38, ERK pathways. Exerts anti-hypertrophic effects. Caspase-1 activity inhibitor in vivo and in vitro. Antibacterial and antilisteria compound. L-Type calcium channel blocker.
Catalog No. 50-205-3442