Learn More
MSC Imperial Swivel Handle Tube Bender, 1/4 in O.D. Tubing, 180 Degree Bends
Catalog No. 5000508060
Encompass
This swivel handle tube bender is designed for precision bending of 1/4 in O.D. tubing. Its unique swivel handle mechanism allows for repositioning mid-bend, enabling effortless fabrication of smooth, tight radius bends up to 180 degrees while reducing effort, improving control, and preventing handle crossing.
  • Swivel handle repositions for leverage during bends.
  • Fabricates smooth, 180-degree bends with reduced effort.
  • Eliminates handle crossing.
  • Designed for 1/4 in O.D. tubing; each tool bends one size.
  • Open-side design slips over tubing at any point.
  • Prevents marking or flattening of tubing.
  • Suitable for steel, stainless steel, copper, aluminum, and other metal tubing.
  • Conforms to Federal Specification GGG-B-191a Type III, Class 2.
  • Calibrated to show bend angle.
Catalog No. 50-005-08060 Supplier MSC Supplier No. 56555295
This swivel handle tube bender is designed for precision bending of 1/4 in O.D. tubing. Its unique swivel handle mechanism allows for repositioning mid-bend, enabling effortless fabrication of smooth, tight radius bends up to 180 degrees while reducing effort, improving control, and preventing handle crossing.
  • Swivel handle repositions for leverage during bends.
  • Fabricates smooth, 180-degree bends with reduced effort.
  • Eliminates handle crossing.
  • Designed for 1/4 in O.D. tubing; each tool bends one size.
  • Open-side design slips over tubing at any point.
  • Prevents marking or flattening of tubing.
  • Suitable for steel, stainless steel, copper, aluminum, and other metal tubing.
  • Conforms to Federal Specification GGG-B-191a Type III, Class 2.
  • Calibrated to show bend angle.