Learn More
Med Vet International Impervious Drape, "60 x 80", Sterile, J0259B, 5-Pack
Catalog No. 5000582247
Encompass_Preferred
Once you try this drape, you will want it for all your extended procedures! Base material: Sontara laminated with polyethylene film making it completely impervious. This drape has the highest tear strength of any drape fabric. It can be readily fenestrated at the time of surgery without the need for reinforcement. Fanfolded in easy opening header packer. Sterile individual wrap Covers entire patient atient.
Catalog No. 50-005-82247 Supplier Med Vet International Supplier No. JOR259B
Once you try this drape, you will want it for all your extended procedures! Base material: Sontara laminated with polyethylene film making it completely impervious. This drape has the highest tear strength of any drape fabric. It can be readily fenestrated at the time of surgery without the need for reinforcement. Fanfolded in easy opening header packer. Sterile individual wrap Covers entire patient atient.