Learn More
Ace Glass, Inc. Impinger, 125mL, complete with open tube end inlet/outlet, 24/25, 0.035in capillary tip
SDP
Catalog No. 509557838
Encompass_Preferred
125mL high velocity impinge which passes 6L/min when the pressure drop across the orifice is 41cm Hg or greater. Complete with 24/25 impinger stopper with tube end inlet/outlets and a 0.035in capillary tip that is positioned 4mm above the bottom of the bottle. Stopper fits 24/25, 125mL bottle and features a 11mm O.D. center tube, and a 8mm O.D. side arm with bulge. Particulate capture range: 0.6 to 2 microns.
Catalog No. 50-955-7838 Supplier Ace Glass, Inc. Supplier No. 754110
125mL high velocity impinge which passes 6L/min when the pressure drop across the orifice is 41cm Hg or greater. Complete with 24/25 impinger stopper with tube end inlet/outlets and a 0.035in capillary tip that is positioned 4mm above the bottom of the bottle. Stopper fits 24/25, 125mL bottle and features a 11mm O.D. center tube, and a 8mm O.D. side arm with bulge. Particulate capture range: 0.6 to 2 microns.