Learn More
Chemglass Life Sciences Midget Impinger, Graduated, Complete, 24/40 Joint Size, Plain Tip
SDP
Catalog No. 50873331
Encompass_Preferred
Midget Impinger, Graduated, Complete, 24/40 Joint Size, Plain Tip. Micro impinger designed for sampling of small volumes of air at low jet velocity, as per EPA Method 6. Impinger bottle is graduated from 5 to 30ml in 5ml subdivisions. Both the bottle and stopper have 24/40 standard taper joints. The orifice at the impinger tip is 1mm, passing 0.09 to 0.11 CFM of air at 12in H20 vacuum. Equipped with plain tubulations.
Catalog No. 50-873-331 Supplier Chemglass Life Sciences Supplier No. CG182001
Midget Impinger, Graduated, Complete, 24/40 Joint Size, Plain Tip. Micro impinger designed for sampling of small volumes of air at low jet velocity, as per EPA Method 6. Impinger bottle is graduated from 5 to 30ml in 5ml subdivisions. Both the bottle and stopper have 24/40 standard taper joints. The orifice at the impinger tip is 1mm, passing 0.09 to 0.11 CFM of air at 12in H20 vacuum. Equipped with plain tubulations.