Learn More
Chemglass Life Sciences Midget Impinger, Plain Top, Fritted Tube, Graduated, Complete
SDP
Catalog No. 50873332
Encompass_Preferred
Midget Impinger, Plain Top, Fritted Tube, Graduated, Complete. Micro impinger designed for sampling of small volumes of air at low jet velocity, as per EPA Method 6. Impinger bottle is graduated from 5 to 30mL in 5mL subdivisions. Both the bottle and stopper have 24/40 standard taper joints. The nozzle being replaced with an extra coarse fritted gas dispersion tube which enhances absorption by trapping the liquid.
Catalog No. 50-873-332 Supplier Chemglass Life Sciences Supplier No. CG182101
Midget Impinger, Plain Top, Fritted Tube, Graduated, Complete. Micro impinger designed for sampling of small volumes of air at low jet velocity, as per EPA Method 6. Impinger bottle is graduated from 5 to 30mL in 5mL subdivisions. Both the bottle and stopper have 24/40 standard taper joints. The nozzle being replaced with an extra coarse fritted gas dispersion tube which enhances absorption by trapping the liquid.