Learn More
IMPLEN INC Implen N120-Mobile Nano Photometer, 12-channel Mirovolume (drop) UV/Vis Spectrophtometer with built-in battery pack
Supplier:  IMPLEN INC N120MOBILE
Encompass_Preferred
Implen is a leader in innovative, high-quality UV/VIS spectroscopy, and the NanoPhotometer trusted by thousands of labs worldwide. The NanoPhotometer N120 scans 12 drops per run from 200–900 nm in 20s for applications including quantification of DNA, RNA, proteins, kinetics, nanoparticles, and antibodies using only 2 µl drops. A recalibration-free design removes the need for costly service contracts, guaranteeing accuracy and stability without pedestal reconditioning. Advanced measurement technology eliminates innaccuracies from low surface tension and evaporation, ensuring reproducible results even with proteins or organic solvents. Sample Control™ and Blank Control™ enhance contaminant detection via ratios and spectral analysis, improving success of downstream applications such as (q)PCR, cloning, microarrays, drug production, and protein purification. Optional CFR21 software, IQ/OQ package, and REST API for LIMS integration make the N120 ideal for pharmaceutical labs
Catalog No. NC3198809
Add to Cart
Request a Quote