Learn More
TARGETMOL CHEMICALS INC IMPORTAZOLE 25MG
SDP
Catalog No. 502580260
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Importazole is an inhibitor of the nuclear transport receptor importin-(beta). purity: 99%
Catalog No. 50-258-0260 Supplier TARGETMOL CHEMICALS INC Supplier No. T344525MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Importazole is an inhibitor of the nuclear transport receptor importin-(beta). purity: 99%