Learn More
Abcam Importazole, nuclear transport receptor importin-beta inhibitor, 5MG
Catalog No. 502704911
Encompass_Preferred
MW 318.42 g/mol, Purity >99%. Selective nuclear transport receptor importin-β inhibitor (IC₅₀ = 25 μM). Blocks SINC localization at the nuclear envelope. Impairs release of importin-β cargo and induces defects in defects in spindle assembly during mitosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4911 Supplier Abcam Supplier No. AB1461555MG
Add to Cart
Add to Cart
MW 318.42 g/mol, Purity >99%. Selective nuclear transport receptor importin-β inhibitor (IC₅₀ = 25 μM). Blocks SINC localization at the nuclear envelope. Impairs release of importin-β cargo and induces defects in defects in spindle assembly during mitosis.
The product is subject to the following: Abcam Restricted Use Statement