Learn More
Mopec Impression Foam
SDP
Catalog No. 22446142 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W x H):
14 x 6 x 2.25 in.
14 x 12 x 2.25 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
22-446-142 14 x 12 x 2.25 in.
22-446-093 14 x 6 x 2.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-142 Supplier Mopec Supplier No. FS171
Add to Cart
Add to Cart
Impression foam designed to capture detailed three dimensional impressions of footprints, footwear, and other surface evidence.
  • Easy-close lid
  • Built-in storage for impressions
  • Provides an easy, clean, and efficient way to obtain an impression of suspected footwear
  • Made from sturdy cardboard and foam.
  • Soft, high resolution casting material used in forensic science
  • Used to capture fine surface features, preserving ridge details and tread patterns for accurate forensic analysis.
Specifications
Product Type Impression Foam
Color Blue
Dimensions (L x W x H) 14 x 12 x 2.25 in.