Learn More
Biocare Medical, LLC IMPROV REAGENT VIAL 15ML
SDP
Catalog No. NC2585312
Encompass
The ONCORE™ Pro Improv Reagent Vials are “user-fillable” vials that are designed for the utilization of alternate primary antibodies not offered within the ONCORE reagent line on the ONCORE Automated Slide Staining System.
Catalog No. NC2585312 Supplier Biocare Medical, LLC Supplier No. ONCPR102JJ
The ONCORE™ Pro Improv Reagent Vials are “user-fillable” vials that are designed for the utilization of alternate primary antibodies not offered within the ONCORE reagent line on the ONCORE Automated Slide Staining System.