Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Improve Medical IMPROVACUTER Blood Collection Tubes

Catalog No. p-7227828
Click to view available options
Capacity (Metric):
5 mL
8 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

FDA-approved, IMPROVACUTER Shatterproof PET tubes offer a safe method of blood collection and reduce the potential for tube breakage and specimen spillage, thereby reducing the potential for exposure to bloodborne pathogens.

  • These FDA-approved, shatterproof PET tubes offer a safe method of blood collection
  • They reduce the potential for tube breakage and specimen spillage, thereby reducing the potential for exposure to bloodborne pathogens
  • IMPROVACUTER Gel and Clot Activator Tubes offer serum for determinations in chemistry and serology
  • The tube contains a gel that forms a stable barrier between the serum and the cellular components of the whole blood after centrifugation
  • They are coated with silicone and micronized silica particles to accelerate clotting and reduce the adherence of red cells to tube walls
  • The safety closure has a unique rubber stopper that seals IMPROVACUTER blood collection tubes
  • It is recessed within and covered by a plastic shield to help protect laboratory personnel from contact with blood on the stopper or around the outer rim of the tube
  • The closure also helps prevent blood from splattering when the tube is opened

Specifications

Color Clear
Material PET
Sterility Sterile
Closure Type Safety Pull Cap
Capacity (Metric) 5 mL
Diameter (Metric) 13 mm
Length (Metric) 100 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.