Learn More
Electron Microscopy Sciences Improved Neubauer Double Cell 0.1 mm Haemocytometer
SDP
Catalog No. 50366666
Encompass_Preferred
Hemocytometer Counting cell is used for hemoglobin and leucocyte. Double Cell 0.1mm, Ruling allows full use central squares
Catalog No. 50-366-666 Supplier Electron Microscopy Sciences Supplier No. 6805216
Hemocytometer Counting cell is used for hemoglobin and leucocyte. Double Cell 0.1mm, Ruling allows full use central squares