Learn More
MSC Value Collection Impulse Sealer, 20 in Max Seal, 1 kit
Catalog No. 501918425
Encompass
This impulse sealer service kit is designed for units with a maximum seal size of 20 in, providing essential replacement components to maintain efficient operation of your sealing equipment.
  • Includes one PTFE tape strip
  • Includes one PTFE cover
  • Includes two heating elements
  • Maximum seal size of 20 in
  • Country of origin: Taiwan
Catalog No. 50-191-8425 Supplier MSC Supplier No. 39580154
This impulse sealer service kit is designed for units with a maximum seal size of 20 in, providing essential replacement components to maintain efficient operation of your sealing equipment.
  • Includes one PTFE tape strip
  • Includes one PTFE cover
  • Includes two heating elements
  • Maximum seal size of 20 in
  • Country of origin: Taiwan