Learn More
Medchemexpress LLC Impurity C of alfacalcidol | 82266-85-1 | MFCD22124488 | 99.8% | C35H49N3O4 | 10MG
SDP
Catalog No. 5000258479
Encompass_Preferred
Impurity C of alfacalcidol is an analytical reference impurity of the vitamin D analogue alfacalcidol, supplied as a high-purity solid for research use. It is intended for impurity profiling, analytical method development, and reference standard applications; follow recommended storage to preserve stability.
  • High reported purity of 99.8%.
  • Available in small solid quantities including 10 mg.
  • Chemical formula C35H49N3O4; molecular weight 575.78.
  • Suitable for impurity profiling and method development.
  • Store protected from light and under inert atmosphere; in solvent keep at -80°C for long term.
Catalog No. 50-002-58479 Supplier Medchemexpress LLC Supplier No. HY1329410MG
Impurity C of alfacalcidol is an analytical reference impurity of the vitamin D analogue alfacalcidol, supplied as a high-purity solid for research use. It is intended for impurity profiling, analytical method development, and reference standard applications; follow recommended storage to preserve stability.
  • High reported purity of 99.8%.
  • Available in small solid quantities including 10 mg.
  • Chemical formula C35H49N3O4; molecular weight 575.78.
  • Suitable for impurity profiling and method development.
  • Store protected from light and under inert atmosphere; in solvent keep at -80°C for long term.