Learn More
Medchemexpress LLC Impurity C of calcitriol | 86307-44-0 | MFCD22124487 | 99.8% | C35H49N3O5 | 10MG
SDP
Catalog No. 5000259723
Encompass_Preferred
Impurity C of calcitriol (CAS 86307-44-0) is a characterized impurity of calcitriol used as an analytical reference standard in research and method development. Supplied as a high-purity solid, it is intended for chromatographic and spectroscopic applications; users should consult the lot-specific certificate of analysis for exact specifications.
  • High purity suitable for analytical standards and reference use.
  • Provided as a solid for convenient handling and storage.
  • Compatible with chromatographic and spectroscopic methods.
  • Useful for impurity profiling and method validation.
  • Batch-specific certificate of analysis available for exact specifications.
Catalog No. 50-002-59723 Supplier Medchemexpress LLC Supplier No. HY1329310MG
Impurity C of calcitriol (CAS 86307-44-0) is a characterized impurity of calcitriol used as an analytical reference standard in research and method development. Supplied as a high-purity solid, it is intended for chromatographic and spectroscopic applications; users should consult the lot-specific certificate of analysis for exact specifications.
  • High purity suitable for analytical standards and reference use.
  • Provided as a solid for convenient handling and storage.
  • Compatible with chromatographic and spectroscopic methods.
  • Useful for impurity profiling and method validation.
  • Batch-specific certificate of analysis available for exact specifications.