Learn More
Bachem (Des-Gly10,D-Ser(tBu)6,Pro-NHNH29)-LHRH Trifluoroacetate
SDP
Supplier:  Bachem 4039400.0005
Encompass_Preferred
(Des-Gly10,D-Ser(tBu)6,Pro-NHNH29)-LHRH Trifluoroacetate, Pyr-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-NHNH2, 147688-42-4, C58H83N17O13, Mr 1226.40, 5mg. Decarbamoylgoserelin, (Pro-NHNH29)-Buserelin, (Des-carboxamide)-Goserelin. Impurity E of the Goserelin Ph. Eur. monograph.
Catalog No. 50-194-6709