Learn More
Bachem Pyr-D-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-Azagly-NH2
SDP
Supplier:  Bachem 4035303.0005
Encompass_Preferred
(D-His2,D-Ser(tBu)6,Azagly10)-LHRH , Pyr-D-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-Azagly-NH2, 1926163-37-2, C59H84N18O14, Mr 1269.43, 5mg. (D-His2)-Goserelin. Impurity G of the Goserelin Ph. Eur. monograph.
Catalog No. 50-194-6711