Learn More
Bachem Pyr-His-Trp-Ser(Ac)-Tyr-D-Ser(tBu)-Leu-Arg-Pro-Azagly-NH2
SDP
Supplier:  Bachem 4054732.0005
Encompass_Preferred
(Ser(Ac)4,D-Ser(tBu)6,Azagly10)-LHRH Trifluoroacetate, Pyr-His-Trp-Ser(Ac)-Tyr-D-Ser(tBu)-Leu-Arg-Pro-Azagly-NH2, 1926163-83-8, C61H86N18O15, Mr 1311.47, 5mg. (Ser(Ac)4)-Goserelin. Impurity K of the Goserelin Ph. Eur. monograph.
Catalog No. 50-194-6712