Learn More
Bachem Pyr-His-Trp-Ser-Tyr-D-Ser(tBu)-D-Leu-Arg-Pro-Azagly-NH2
SDP
Supplier:  Bachem 4037082.0005
Encompass_Preferred
(D-Ser(tBu)6,D-Leu7,Azagly10)-LHRH, Pyr-His-Trp-Ser-Tyr-D-Ser(tBu)-D-Leu-Arg-Pro-Azagly-NH2, 1926163-38-3, C59H84N18O14, Mr 1269.43, 5mg. (D-Leu7)-Goserelin. Impurity L of the Goserelin Ph. Eur. monograph.
Catalog No. 50-194-6716