Learn More
Sigma Aldrich Fine Chemicals Biosciences IMR-1 >=98% (HPLC) | 310456-65-6 | 5MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SML18125MG
Encompass
IMR-1 >=98% (HPLC) | Purity: >=98% (HPLC) | Mol Wt: 353.41 | 310456-65-6 | 5MG
Catalog No. 50-186-6077