Learn More
ATCC IMR-90 Lung Fibroblast Human (Homo sapiens)
Catalog No. 502384913
Encompass
The IMR-90 cell line is made up of fibroblasts isolated from normal lung tissue derived from a 16-week-old female. IMR-90 cells are reported to be capable of attaining 58 population doublings before the onset of senescence. The cell line may be used as an alternate to WI-38 and other human lung cell strains, considering division potential and viral susceptibilities.
Catalog No. 50-238-4913 Supplier ATCC Supplier No. CCL186
The IMR-90 cell line is made up of fibroblasts isolated from normal lung tissue derived from a 16-week-old female. IMR-90 cells are reported to be capable of attaining 58 population doublings before the onset of senescence. The cell line may be used as an alternate to WI-38 and other human lung cell strains, considering division potential and viral susceptibilities.