Learn More
Medchemexpress LLC HY-114200 100mg Medchemexpress, Imrecoxib CAS:395683-14-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114200100MG
Encompass_Preferred
Medchemexpress, HY-114200 100mg Imrecoxib CAS:395683-14-4 Imrecoxib (BAP-909) is a novel and selective cyclooxygenase 2 ( COX-2 ) inhibitor with an IC 50 value of 18 nM, it also inhibits COX1- activity with an IC 50 value of 115 nM. Imrecoxib (BAP-909) has anti-inflammatory effect [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2471