Learn More
Medchemexpress LLC HY-134539 5mg Medchemexpress, IMT1 CAS:2304621-31-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1345395MG
Encompass_Preferred
Medchemexpress, HY-134539 5mg IMT1 CAS:2304621-31-4 IMT1 is a first-in-class specific and noncompetitive human mitochondrial RNA polymerase (POLRMT) inhibitor. IMT1 causes a conformational change of POLRMT, which blocks substrate binding and transcription in a dose-dependent way in vitro. IMT1 reduces deoxynucleoside triphosphate levels and citric acid cycle intermediates, resulting in a marked depletion of cellular amino acid levels. IMT1 has the potential for mitochondrial transcription disorders related diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1369