Learn More
Medchemexpress LLC HY-137067 5mg , IMT1B CAS:2304621-06-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1370675MG
Encompass_Preferred
HY-137067 5mg IMT1B CAS: 2304621-06-3 IMT1B (LDC203974) is an orally active, noncompetitive and specific allosteric inhibitor of mitochondrial RNA polymerase (POLRMT) and inhibits mitochondrial DNA (mtDNA) expression. IMT1B has anti-tumour effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9064